Learn More
UCHL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533098
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Mouse, Rat, Dog, Elephant, Opossum, Turkey, Chicken, Lizard (100%); Orangutan, Monkey, Bovine, Horse, Rabbit, Pig, Salmon (94%); Pufferfish, Zebrafish, Stickleback, Water flea (89%); Xenopus (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
UCHL3 Polyclonal antibody specifically detects UCHL3 in Chicken, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
UCHL3 | |
Polyclonal | |
Unconjugated | |
UCHL3 | |
RGD1561196; testicular tissue protein Li 221; ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L3; ubiquitin C-terminal hydrolase L3; Ubiquitin thioesterase L3; ubiquitin thiolesterase; UCHL3; UCH-L3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
395626, 498560, 50933, 7347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P15374, Q91Y78, Q9JKB1 | |
UCHL3 | |
Synthetic 18 amino acid peptide from internal region of human UCHL3. | |
50 μg | |
Primary | |
Chicken, Human, Mouse, Rat | |
Antibody | |
IgG |