missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1187
Code : NEW
Additional Details : Weight : 0.00900kg
Description
PA1-187 detects both the ubiquitin monomer as well as poly-ubiquitinated proteins of various molecular weights.
Ubiquitin Polyclonal antibody specifically detects Ubiquitin in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Specifications
Ubiquitin | |
Polyclonal | |
Unconjugated | |
UBB | |
0610006J14Rik; 1810034D23Rik; 2700054O04Rik; 40S ribosomal protein S27a; 40S ribosomal protein S27-like; 40S ribosomal protein S27-like protein; AI194771; AL033289; CEP80; CG5271; CG5271-PA; Chap1; Dmel\CG5271; Dmel_CG5271; Dub80; epididymis luminal protein 112; epididymis secretory protein Li 50; HEL112; HEL-S-50; hm:zeh0386; HMG20; hPLIC-2; HRIHFB2157; HUBCEP52; ik:tdsubc_1f2; l(2)04820; l(2)31Ei; LOC100912032; LOC101747587; male female sterile 31; mfs(2)31; mfs(2)48; mfs48; OTTHUMP00000200786; OTTHUMP00000201692; PLIC-2; polyubiquitin; polyubiquitin B; polyubiquitin C; poly-ubiquitin precursor; polyubiquitin-B; Polyubiquitin-C; polyubiquitin-C-like; RE44350p; ribosomal protein S27; ribosomal protein S27 like; ribosomal protein S27A; ribosomal protein S27a L homeolog; ribosomal protein S27a S homeolog; ribosomal protein S27-like; ribosomal protein S27-like protein; RPL40; RpS27; Rps27a; rps27a.L; rps27a.S; RpS27A-PA; RPS27L; S27A; similar to polyubiquitin; similar to ubiquitin-S27a f | |
Rabbit | |
Protein G | |
RUO | |
100912032, 192255, 22186, 22187, 22190, 403722, 474599, 479513, 50522, 51065, 610457, 610569, 6233, 64156, 67941, 681429, 7311, 7314, 7316, 78294 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P0CG47, P0CG48, P0CG49, P0CG50, P0CG51, P24051, P62979, P62982, P62983, P62984, P62986, P62987, P63050, Q63429, Q6ZWY3, Q71UM5 | |
RPS27A, RPS27L, UBA52, UBB, UBC | |
Full-length monoubiqutin | |
100 μg | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |