missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA126088
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-26088 detects Ubiquitin from human and mouse samples.
Ubiquitin Polyclonal antibody specifically detects Ubiquitin in Bovine, Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Ubiquitin | |
Polyclonal | |
Unconjugated | |
UBB | |
0610006J14Rik; 1810034D23Rik; 2700054O04Rik; 40S ribosomal protein S27a; 40S ribosomal protein S27-like; 40S ribosomal protein S27-like protein; AI194771; AL033289; CEP80; CG5271; CG5271-PA; Chap1; Dmel\CG5271; Dmel_CG5271; Dub80; epididymis luminal protein 112; epididymis secretory protein Li 50; HEL112; HEL-S-50; hm:zeh0386; HMG20; hPLIC-2; HRIHFB2157; HUBCEP52; ik:tdsubc_1f2; l(2)04820; l(2)31Ei; LOC100912032; LOC101747587; male female sterile 31; mfs(2)31; mfs(2)48; mfs48; OTTHUMP00000200786; OTTHUMP00000201692; PLIC-2; polyubiquitin; polyubiquitin B; polyubiquitin C; poly-ubiquitin precursor; polyubiquitin-B; Polyubiquitin-C; polyubiquitin-C-like; RE44350p; ribosomal protein S27; ribosomal protein S27 like; ribosomal protein S27A; ribosomal protein S27a L homeolog; ribosomal protein S27a S homeolog; ribosomal protein S27-like; ribosomal protein S27-like protein; RPL40; RpS27; Rps27a; rps27a.L; rps27a.S; RpS27A-PA; RPS27L; S27A; similar to polyubiquitin; similar to ubiquitin-S27a f | |
Rabbit | |
Protein A | |
RUO | |
22186, 22187, 22190, 281370, 286839, 444874, 51065, 614220, 615199, 6233, 67941, 7311, 7314, 7316, 78294 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2 | |
P0CG47, P0CG48, P0CG49, P0CG50, P0CG53, P0CH28, P62979, P62983, P62984, P62987, P62992, P63048, Q3T0B7, Q6ZWY3, Q71UM5 | |
RPS27A, RPS27L, UBA52, UBB, UBC | |
Full length native Ubiquitin protein purified from bovine red blood cells. | |
100 μL | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG |