missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531698
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (82%), Rat (82%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UBAC1 Polyclonal antibody specifically detects UBAC1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
UBAC1 | |
Polyclonal | |
Unconjugated | |
UBAC1 | |
1110033G07Rik; AA407978; E3 ubiquitin-protein ligase subunit KPC2; GBDR1; Gdbr1; glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; KPC2; putative glialblastoma cell differentiation-related; putative glialblastoma cell differentiation-related protein; RP11-432J22.3; testicular secretory protein Li 64; UBA domain containing 1; UBA domain-containing protein 1; Ubac1; Ubadc1; ubiquitin associated domain containing 1; ubiquitin-associated domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10422 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BSL1 | |
UBAC1 | |
Recombinant fragment corresponding to a region within amino acids 69 and 348 of Human UBAC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |