missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF1L4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536004
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Human (99%).
U2AF1L4 Polyclonal antibody specifically detects U2AF1L4 in Human samples. It is validated for Immunocytochemistry.Specifications
U2AF1L4 | |
Polyclonal | |
Unconjugated | |
U2AF1L4 | |
FLJ35525; MGC33901; Splicing factor U2AF 26 kDa subunit; U2 auxiliary factor 26; U2 small nuclear RNA auxiliary factor 1 like 4; U2 small nuclear RNA auxiliary factor 1-like 3; U2 small nuclear RNA auxiliary factor 1-like 4; U2 small nuclear RNA auxiliary factor 1-like protein 3; U2 small nuclear RNA auxiliary factor 1-like protein 4; U2(RNU2) small nuclear RNA auxiliary factor 1-like 3; U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3; U2AF1L3; U2AF1L3V1; U2AF1L4; U2AF1-like 4; U2AF1-like protein 3; U2AF1RS3; U2AF1-RS3; U2af26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
199746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8WU68 | |
U2AF1L4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 202 of U2AF1L4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |