missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen SA510033
Code : PN
Additional Details : Weight : 0.01000kg
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, horse, human, mouse, pig and rat.
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 550 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Rabbit IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |