missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21430
164.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Description
Product will be shipped at Room Temperature.
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 555 | |
Goat | |
purified | |
RUO | |
Rabbit | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |