missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-WEE1 (Ser642) Monoclonal Antibody (T.870.10), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514806
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Phospho-WEE1 (Ser642) Monoclonal antibody specifically detects Phospho-WEE1 (Ser642) in Bovine, Human, Mouse, Non-human Primate, Rat, X. laevis, Zebrafish samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Phospho-WEE1 (Ser642) | |
Monoclonal | |
Unconjugated | |
WEE1 | |
CTRCT30; DKFZp686I18166; epididymis luminal protein 113; fa20h03; FLJ16446; HEL113; VIM; Vimentin; wee 1 homolog; WEE 1 homolog 1 (S. pombe); Wee1; WEE1 G2 checkpoint kinase; WEE1 homolog; wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; wee1A kinase; WEE1hu; wee1-like protein kinase; wu:fa20h03; zgc:136514 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22390, 308937, 327060, 540322, 7465 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
T.870.10 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P30291, P47810, Q63802 | |
WEE1 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer642 of human wee1 | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rat, X. laevis, Zebrafish | |
Antibody | |
IgG |