missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AMPK beta-1 (Ser181) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538405
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Non-catalytic subunit of AMP-activated protein kinase (AMPK), an energy sensor protein kinase that plays a key role in regulating cellular energy metabolism. In response to reduction of intracellular ATP levels, AMPK activates energy-producing pathways and inhibits energy-consuming processes: inhibits protein, carbohydrate and lipid biosynthesis, as well as cell growth and proliferation. AMPK acts via direct phosphorylation of metabolic enzymes, and by longer-term effects via phosphorylation of transcription regulators. Also acts as a regulator of cellular polarity by remodeling the actin cytoskeleton; probably by indirectly activating myosin. Beta non-catalytic subunit acts as a scaffold on which the AMPK complex assembles, via its C-terminus that bridges alpha (PRKAA1 or PRKAA2) and gamma subunits (PRKAG1, PRKAG2 or PRKAG3).Specifications
Phospho-AMPK beta-1 (Ser181) | |
Polyclonal | |
Unconjugated | |
PRKAB1 | |
beta-1 subunit, AMPK beta-1 chain, AMPKb, pAMPK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19079, 5564, 83803 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y478, Q9R078, P80386 | |
PRKAB1 | |
A synthetic phosphopeptide derived from human AMPK beta 1 around the phosphorylation site of Ser181 (S-S-SP-P-P) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |