missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTSR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA141575
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: brain lysate, human brain protein.
NTSR2 Polyclonal antibody specifically detects NTSR2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NTSR2 | |
Polyclonal | |
Unconjugated | |
NTSR2 | |
High-affinity levocabastine-sensitive neurotensin receptor; levocabastine-sensitive neurotensin receptor; low affinity neurotensin receptor; low-affinity levocabastine-sensitive neurotensin receptor; neurotensin receptor 2; neurotensin receptor type 2; neurotensin receptor, type 2; NT2 receptor; NT2R; NTR2; NT-R-2; NTR2 receptor; NTRL; Ntsr2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18217, 23620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
O95665, P70310 | |
NTSR2 | |
Synthetic peptide corresponding to residues 150-200 of human Neurotensin receptor 2 . | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |