Learn More
NF1 Monoclonal Antibody (McNFn27b), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1085
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-085 detects neurofibromin in human, rat, and mouse cells. MA1-085 has been successfully used in Western blot, and immunohistochemistry procedures. By Western blot, this antibody detects a ∽240-260 kDa protein. The MA1-085 immunogen is a synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.
NF1 Monoclonal antibody specifically detects NF1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
NF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P21359, P97526, Q04690 | |
NF1 | |
Synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin. | |
200 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
McNFn27b | |
Unconjugated | |
NF1 | |
AW494271; DKFZp686J1293; E030030H24Rik; FLJ21220; neurofibromatosis 1; Neurofibromatosis type 1; neurofibromatosis-related protein NF-1; Neurofibromin; neurofibromin 1; Neurofibromin truncated; NF1; Nf-1; NFNS; VRNF; WSS | |
Mouse | |
Protein G | |
RUO | |
18015, 24592, 4763 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |