missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEURL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539285
Code : NEW
Additional Details : Weight : 0.00900kg
Description
NEURL is involved in the determination of cell fate in the neurogenic region of the embryo and plays a role in the determination of cell fate in the central nervous system. NEURL may act as a tumor suppressor whose inactivation could be associated with maligt progression of astrocytic tumors.Specifications
NEURL1 | |
Polyclonal | |
Unconjugated | |
NEURL1 | |
Neuralized like protein 1, h neuralized 1, h neu, RING finger protein 67, NEURL1, RNF67, Neuralized homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9148 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O76050 | |
NEURL1 | |
A synthetic peptide derived from the internal region of human NEURL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |