missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEIL1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518134
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and porcine based on sequence homology.
NEIL1 Polyclonal antibody specifically detects NEIL1 in Human samples. It is validated for Immunohistochemistry.Specifications
NEIL1 | |
Polyclonal | |
Unconjugated | |
NEIL1 | |
2810450N13Rik; DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; endonuclease 8-like 1; endonuclease VIII; endonuclease VIII-like 1; FLJ22402; FPG1; hFPG1; NEH1; nei endonuclease VIII-like 1; nei endonuclease VIII-like 1 (E. coli); nei homolog 1; nei like DNA glycosylase 1; Nei1; NEIL1; nei-like DNA glycosylase 1; nei-like protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
79661 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96FI4 | |
NEIL1 | |
Synthetic peptide sequence (DIPSLEPEGTSAS) corresponding to the C-terminus amino acids of NEIL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |