Learn More
NBR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530085
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, NIH-3T3, JC. Predicted reactivity: Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NBR1 Polyclonal antibody specifically detects NBR1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
NBR1 | |
Polyclonal | |
Unconjugated | |
NBR1 | |
1A13B; 1A1-3B; B-box protein; Ca125; Cell migration-inducing gene 19 protein; IAI3B; KIAA0049; M17S2; Membrane component chromosome 17 surface marker 2; Membrane component chromosome 17 surface marker 2 homolog; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); MIG19; migration-inducing protein 19; mKIAA0049; Nbr1; NBR1, autophagy cargo receptor; neighbor of Brca1 gene 1; neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; next to the Brca1; ovarian carcinoma antigen CA125; Protein 1A1-3B; RGD1311421 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17966, 4077 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P97432, Q14596 | |
NBR1 | |
Recombinant protein fragment corresponding to a region within amino acids 389 and 678 of Human NBR1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |