Learn More
NAA16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531688
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (96%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NAA16 Polyclonal antibody specifically detects NAA16 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NAA16 | |
Polyclonal | |
Unconjugated | |
NAA16 | |
1300019C06Rik; N(alpha)-acetyltransferase 16, NatA auxiliary subunit; NAA16; Nalpha acetyltransferase 16; N-alpha-acetyltransferase 16, NatA auxiliary subunit; Narg1l; NARG1-like protein; NAT2; NMDA receptor regulated 1-like protein; NMDA receptor-regulated 1-like protein; NMDA receptor-regulated 1-like protein-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6N069 | |
NAA16 | |
Recombinant fragment corresponding to a region within amino acids 1 and 187 of Human NARG1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |