Learn More
CRALBP Monoclonal Antibody (B2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1813
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-813 detects CRALBP protein in bovine, human, mouse, non-human primate and rat samples. MA1-813 has successfully been used in Western blot, immunohistochemistry, immunoprecipitation, and immunofluorescence procedures. By Western blot, this antibody detects a 37 kDa protein representing human recombinant CRALBP. The MA1-813 immunogen is purified human recombinant CRALBP.
CRALBP Monoclonal antibody specifically detects CRALBP in Bovine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CRALBP | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P10123, P12271, Q9Z275 | |
RLBP1 | |
Human recombinant CRALBP. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
B2 | |
Unconjugated | |
RLBP1 | |
3110056M11Rik; cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; retinaldehyde binding protein 1; retinaldehyde-binding protein 1; retinaldehyde-binding protein precursor; Rlbp1 | |
Mouse | |
Protein A | |
RUO | |
19771, 282038, 293049, 6017 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |