Learn More
Claudin 4 Monoclonal Antibody (3E2C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 329400
352.28 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02100kg
Description
Reactivity has been confirmed with human, rat ileum homogenates, and canine MDCK cell lysates by western blotting and immunofluorescence. Reactivity has also been confirmed with formalin-fixed, paraffin-embedded (FFPE) human normal colon and colon cancer tissues by immunohistochemistry. For best results in immunohistochemistry with formalin-fixed, paraffin-embedded (FFPE) tissues, heat induced epitope retrieval (HIER) with citrate buffer, pH 6.0, is required prior to staining. This antibody reacts specifically with the ∽ 22 kDa Claudin-4 protein and does not cross-react with the protein at 55 kDa.
Claudin 4 Monoclonal antibody specifically detects Claudin 4 in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Claudin 4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
O14493, O35054 | |
Cldn4 | |
Synthetic peptide corresponding to a 22 amino acid sequence derived from the C-terminal region of human Claudin-4. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
3E2C1 | |
Unconjugated | |
Cldn4 | |
Cep-r; claudin 4; CLAUDIN4; claudin-4; CLDN4; Clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPER; CPE-R; CPE-receptor; Cpetr; CPETR1; h CPE-receptor; hCPE-R; WBSCR8; Williams-Beuren syndrome chromosomal region 8 protein | |
Mouse | |
Protein A | |
RUO | |
100856416, 12740, 1364, 304407 | |
-20°C | |
Liquid |