missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Amylase Monoclonal Antibody (AMY10-108.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110620
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-10620 detects Amylase from human samples. MA1-10620 has been successfully used in ELISA applications. The MA1-10620 immunogen is purified human pancreatic Amylase.
Pancreatic Amylase Monoclonal antibody specifically detects Pancreatic Amylase in Human samples. It is validated for ELISA.Specifications
Pancreatic Amylase | |
Monoclonal | |
0.3 mg/mL | |
PBS with no preservative; pH 7.4 | |
P04746, P19961 | |
AMY2A, AMY2B | |
Purified human pancreatic Amylase. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgM |
ELISA | |
AMY10-108.5 | |
Unconjugated | |
AMY2B | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A | |
Mouse | |
Protein G | |
RUO | |
279, 280 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |