missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Amylase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525330
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with porcine based on sequence homology.
Pancreatic Amylase Polyclonal antibody specifically detects Pancreatic Amylase in Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Pancreatic Amylase | |
Polyclonal | |
Unconjugated | |
AMY2A | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100043207, 100043684, 279, 280, 545562 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P04746, P19961 | |
AMY2A, Amy2a1, Amy2a4, AMY2B | |
KLH conjugated synthetic peptide between 484-511 amino acids from the C-terminal region of human AMY2A | |
400 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |