Learn More
AMPK gamma-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527471
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mouse liver. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (88%), Xenopus laevis (80%), Pig (100%), Chicken (92%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AMPK gamma-1 Polyclonal antibody specifically detects AMPK gamma-1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.Specifications
AMPK gamma-1 | |
Polyclonal | |
Unconjugated | |
PRKAG1 | |
5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase, gamma-1 subunit; AA571379; AMP-activated protein kinase, noncatalytic gamma-1 subunit; ampk gamma; AMPK gamma1; AMPK gamma-1 chain; AMPK subunit gamma-1; AMPKG; BB036179; Prkaac; PRKAG1; Prkga1; Protein kinase AMP-activated gamma; protein kinase AMP-activated non-catalytic subunit gamma 1; protein kinase, AMP-activated, gamma 1 non-catalytic subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19082, 5571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O54950, P54619 | |
PRKAG1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 224 of Human AMPK gamma 1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |