missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMICA Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA548027
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human AMICA is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
AMICA Polyclonal antibody specifically detects AMICA in Mouse samples. It is validated for Western Blot.Specifications
AMICA | |
Polyclonal | |
Unconjugated | |
JAML | |
adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
270152 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q80UL9 | |
JAML | |
Mouse myeloma cell line NS0-derived recombinant mouse AMICA/JAML Leu23-Leu281 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |