Learn More
ALPPL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542222
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SESGSPEYRQ QSAVPLDGET HAGEDVAVFA RGPQAHLVHG VQEQTFIAHV Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 79%; Zebrafish: 86%
ALPPL2 Polyclonal antibody specifically detects ALPPL2 in Human samples. It is validated for Western Blot.Specifications
ALPPL2 | |
Polyclonal | |
Unconjugated | |
ALPG | |
Akp5; alkaline phosphatase 5; Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; alkaline phosphatase, placental like 2; alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; Alpg; ALPPL; ALPPL2; C77216; D1Ertd816e; EAP; Embryonic alkaline phosphatase; embryonic-type alkaline phosphatase; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; RGD1565100; testicular and thymus alkaline phosphatase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
251 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P10696 | |
ALPG, ALPPL2 | |
Synthetic peptide directed towards the middle region of human ALPPL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |