missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Amylase Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA128946
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-28946 detects alpha Amylase from bacteria samples.
alpha Amylase Polyclonal antibody specifically detects alpha Amylase in Bacteria samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
alpha Amylase | |
Polyclonal | |
HRP | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; a-amylase; alpha-amylase; Alpha-amylase 2B; AMY2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a3; Amy2b; AMY3; amylase 2, pancreatic; amylase 2-2, pancreatic; amylase 2a1; amylase 2a3; amylase 2a3, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; mAmy2-4; mAmy2-6; OTTMUSG00000022430; OTTMUSG00000022432; OTTMUSG00000022462; PA; pancreatic alpha-amylase; pancreatic amylase; pancreatic amylase alpha 2A | |
Alpha Amylase from Bacillus amyloliquefaciens. | |
1 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
10 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |