Learn More
alpha-2c Adrenergic Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
alpha-2c Adrenergic Receptor Polyclonal antibody specifically detects alpha-2c Adrenergic Receptor in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
alpha-2c Adrenergic Receptor | |
Polyclonal | |
Unconjugated | |
ADRA2C | |
[a]2C; Adra 2c; ADRA2C; Adra-2c; ADRA2L2; ADRA2RL2; ADRARL2; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; adrenoceptor alpha 2C; Alpha 2CAR; alpha-2 adrenergic receptor subtype C4; alpha2 AR C4; alpha2 C4; alpha2-AR-C4; alpha2C; Alpha-2C adrenergic receptor; alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha2-C4; alpha-2C-adrenergic receptor; ALPHA2CAR; alpha-2CAR; Alpha-2C-AR; subtype alpha2-C4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.62 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P18825 | |
ADRA2C | |
Synthetic peptide corresponding to a region within amino acids 318 and 409 of Human alpha 2c Adrenergic Receptor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |