missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH5A1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519393
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and canine based on sequence homology.
ALDH5A1 Polyclonal antibody specifically detects ALDH5A1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ALDH5A1 | |
Polyclonal | |
Unconjugated | |
ALDH5A1 | |
6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
214579, 291133, 7915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P51649, P51650, Q8BWF0 | |
ALDH5A1 | |
Synthetic peptide sequence (SQDPAQIWRVAE) corresponding to the internal amino acids of ALDH5A1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |