Learn More
ALDH1A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530288
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MCF-7, mouse testis, rat testis. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (81%), Japanese Medaka (81%), Xenopus laevis (88%), Dog (99%), Chicken (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ALDH1A2 Polyclonal antibody specifically detects ALDH1A2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ALDH1A2 | |
Polyclonal | |
Unconjugated | |
ALDH1A2 | |
AL1A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family member A2; aldehyde dehydrogenase 1 family, member A2; aldehyde dehydrogenase 1A2; aldehyde dehydrogenase family 1 member A2; aldehyde dehydrogenase family 1, subfamily A2; Aldh1a2; Aldh1a7; AV116159; fb50h01; neckless; nls; nof; no-fin; RALDH 2; RALDH(II); Raldh1; RALDH2; Raldh-2; RALDH2-T; retinal dehydrogenase 2; retinal dehydrogenase, type II; retinaldehyde dehydrogenase 2; retinaldehyde dehydrogenase type 2; retinaldehyde-specific dehydrogenase type 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116676, 19378, 8854 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.73 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O94788, Q62148, Q63639 | |
ALDH1A2 | |
Recombinant fragment corresponding to a region within amino acids 2 and 245 of Human ALDH1A2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |