Learn More
CD166 Monoclonal Antibody (5B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517028
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17028 targets AlCAM in IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17028 immunogen is purified recombinant fragment of human AlCAM (amino acids: 405-524) expressed in E. Coli. MA5-17028 detects AlCAM which has a predicted molecular weight of approximately 65.1kDa.
CD166 Monoclonal antibody specifically detects CD166 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD166 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q13740, Q61490 | |
ALCAM | |
Purified recombinant fragment of human AlCAM (amino acids: 405-524) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5B3 | |
Unconjugated | |
ALCAM | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
Mouse | |
Protein G | |
RUO | |
11658, 214 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |