missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMZ2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550330
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total AMZ2 protein.
AMZ2 Polyclonal antibody specifically detects AMZ2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
AMZ2 | |
Polyclonal | |
Unconjugated | |
AMZ2 | |
AA408420; Amz2; archaelysin family metallopeptidase 2; archaemetzincin-2; archaemetzincins-2; archeobacterial metalloproteinase-like 2; archeobacterial metalloproteinase-like protein 2; BM-014; ESTM12; RGD1304846; X83328 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51321 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q86W34 | |
AMZ2 | |
Fusion protein corresponding to residues near the C terminal of human archaelysin family metallopeptidase 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |