missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
30.17 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.
Use promo code "22914" to get your promotional price.
30.17 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Description
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis)
Electrophoresis-grade Ammonium Persulfate is an initiator of acrylamide polymerization for the preparation of polyacrylamide electrophoresis gels.
Specifications
Melting Point | 120°C |
Color | White |
pH | 3.2 |
Physical Form | Powder/Solid |
Quantity | 25 g |
Assay Percent Range | ≥98.0 % |
Linear Formula | (NH4)2S2O8 |
Merck Index | 15, 539 |
Formula Weight | 228.19 |
Percent Purity | ≥98.0% |
Show More |
Safety and Handling
Product Identifier
- Ammonium Persulfate
- Danger
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- H272-May intensify fire; oxidiser.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'