missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™ Total RNA Isolation Kit

Uses glass fiber filter separation technology

£350.00

Specifications

For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Quantity 50 preps
Sample Type Bacteria, Cells, Plant Samples, Tissue, Viral Samples, Yeast
Target Total RNA
View More Specs

Includes: 6mL Formaldehyde Load Dye, 60mL Lysis/Binding Solution, 60mL Water for 64% Ethanol, 4mL Lithium Chloride Precipitation Solution, 40mL Wash Solution 1, 80mL Wash Solution 2/3 Concentrate, 50 Filter Cartridges, 100 Collection Tubes, 10mL Elution Solution

Products 1
Product Code Brand Product Type Includes Price Quantity & Availability  
Product Code Brand Product Type Includes Price Quantity & Availability  
10596935
View accessory products View Documents
Invitrogen™
AM1912M
Kit with manual 6mL Formaldehyde Load Dye; 60mL Lysis/Binding Solution; 60mL Water for 64% Ethanol; 4mL Lithium Chloride Precipitation Solution; 40mL Wash Solution 1; 80mL Wash Solution 2/3 Concentrate; 50 Filter Cartridges; 100 Collection Tubes; 10mL Elution Solution; Manual
£350.00
Each
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RNAqueous Kit is for phenol-free total RNA isolation from 0.1g to 0.5g of tissue or from 107 to 108 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples
How it works
The RNAqueous Kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

Not high-throughput compatible (Manual)

Applications:

DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
50 preps
Bacteria, Cells, Plant Samples, Tissue, Viral Samples, Yeast
Total RNA
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.