missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide Substrate, 10 x 10 x 1mm, Polished One Side, R Plane, Thermo Scientific™
£91.00 - £389.00
Specifications
Chemical Name or Material | Aluminum Oxide Substrate, R Plane |
---|---|
Name Note | Polished Two Sides |
Molecular Formula | Al2O3 |
MDL Number | MFCD00003424 |
Solubility Information | Soluble in water (0.0098g/L at 29°C),hexane and toluene. |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum Oxide Substrate, R Plane | |
Al2O3 | |
Soluble in water (0.0098g/L at 29°C),hexane and toluene. |
Polished Two Sides | |
MFCD00003424 |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures