missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ Aluminum Dewar Cryolabs

Providing state of the art super-insulation and ultra-high vacuum techniques to ensure unrivalled thermal efficiency and vacuum life.

POA

Specifications

Material Aluminum
Includes Neckplugs and dipstick
For Use With (Application) Storage of liquid nitrogen
View More Specs

Description

Description

  • Wide range of capacities available
  • Bucket handles on 10L model
  • Dual handles on 20L model
  • LN2 pressure dispenser for 20L model
  • Strong, lightweight aluminum construction
  • High thermal efficiency
  • Ultra-low evaporative losses
  • 5-year vacuum warranty
  • Ideal for laboratory applications
  • Supplied with neckplugs and dipstick
Specifications

Specifications

Aluminum
Neckplugs and dipstick
Storage of liquid nitrogen
SDS