Learn More
ALPK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA555592
Informations supplémentaires : Weight : 0.02000kg
Description
Immunogen sequence: SPLSSCLPIM THASLGVDTH NSTGQIHDVP ENDIVEPRKR QYVFPVSQKR GTIENERGKP LPSSPDLTRF PCTSSPEGNV TDF Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 51%.
ALPK2 Polyclonal antibody specifically detects ALPK2 in Human samples. It is validated for Immunohistochemistry.Spécification
ALPK2 | |
Polyclonal | |
Unconjugated | |
ALPK2 | |
alpha kinase 2; alpha-kinase 2; alpha-protein kinase 2; Alpk2; Gm549; HAK; heart alpha-kinase; heart alpha-protein kinase; RGD1564431; RNF18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115701 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TB3 | |
ALPK2 | |
Recombinant protein corresponding to Human ALPK2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only