missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha E catenin Mouse anti-Human, Mouse, Clone: 2B7B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66221-1-IG-150UL
This item is not returnable.
View return policy
Description
Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.Specifications
Alpha E catenin | |
Monoclonal | |
2.08 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26231, P35221 | |
CTNNA1 | |
Alpha E catenin Fusion Protein Ag23603 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2B7B8 | |
Unconjugated | |
CTNNA1 | |
Alpha E catenin, Cadherin associated protein, CAP102, Catenin alpha 1, CTNNA1 | |
Mouse | |
Protein G | |
RUO | |
12385, 1495 | |
-20°C | |
Liquid |