Learn More
Thermo Scientific Chemicals ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip
Brand:  Thermo Scientific Chemicals 035235.LQ
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11333607
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
ColorpHast™ pH Strips, Universal, four squares with different indicators on each strip, 0-14 pH range is used in analytical laboratory to measure the pH readings in seconds in drinking water, saliva and in chemical reactions. It is utilized for quick and accurate measurement and saves our time and trouble.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
0 to 6
1 Unit
Intermediate pH Strip