missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15054-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.Specifications
AKR1A1 | |
Polyclonal | |
Unconjugated | |
AKR1A1 | |
AKR1A1, Alcohol dehydrogenase [NADP+], Aldehyde reductase, ALDR1, ALR, ARM, DD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10327, 58810, 78959 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14550, P51635, Q9JII6 | |
Akr1a1 | |
AKR1A1 Fusion Protein Ag7037 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |