missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Adapters for Chromabond™ SPE Columns

For individual packing of SPE columns with Chromabond™ adsorbents. Macherey-Nagel™ Adapters for Chromabond™ SPE Columns are available in several materials for a variety of empty column types and capacities.

£25.50 - £62.00

Specifications

For Use With (Application) Solid Phase Extraction (SPE)
View More Specs

Products 2
Product Code Brand For Use With Material Price Quantity & Availability  
Product Code Brand For Use With Material Price Quantity & Availability  
12471498
View Documents
Macherey-Nagel™
730566
PP columns (30 and 70mL) Polyethylene
£25.50
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
12416646
View Documents
Macherey-Nagel™
730350.4
PP columns (15, 45 and 70mL) Polyethylene This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • PVDF adapter for Glass columns (3 and 6mL)
  • PE adapter for Polypropylene columns (15, 45 and 70mL)
  • PE adapter for Polypropylene columns (30 and 70mL)
Specifications

Specifications

Solid Phase Extraction (SPE)
Videos
SDS
Documents

Documents

Product Certifications