missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals
£32.06 - £925.61
Chemical Identifiers
CAS | 127-08-2 |
---|---|
Molecular Formula | C2H3KO2 |
Molecular Weight (g/mol) | 98.14 |
MDL Number | MFCD00012458 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
IUPAC Name | potassium;acetate |
SMILES | CC(=O)[O-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10328500
|
Thermo Scientific Acros
220151000 |
100 g |
£32.06
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10606042
|
Thermo Scientific Acros
220150010 |
1 kg |
£43.11
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10227363
|
Thermo Scientific Acros
220150050 |
5 kg |
£188.94
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10474985
|
Thermo Scientific Acros
220150250 |
25 kg |
£925.61
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-08-2 | |
98.14 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
Specifications
292.0°C | |
White | |
100.0 | |
99+% | |
CH3CO2K | |
02,96 | |
>250°C | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Pure | |
Plastic bottle |
127-08-2 | |
99.0 | |
Granular Crystalline Powder | |
C2H3KO2 | |
MFCD00012458 | |
01,906; 06,475 | |
15,7726 | |
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
CC(=O)[O-].[K+] | |
98.14 | |
CHEBI:32029 | |
≥99% | |
Authentic | |
Acetic acid, potassium salt, 99+% |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 204-822-2
missing translation for 'rtecsNumber' : AJ3325000
missing translation for 'tsca' : TSCA
RUO – Research Use Only