missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chlorochroman-4-one, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.
Solubility
Soluble in solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.
Solubility
Soluble in solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 101°C to 102°C |
Quantity | 5 g |
Beilstein | 124218 |
Solubility Information | Soluble in solvents. |
Formula Weight | 182.61 |
Percent Purity | 99% |
Chemical Name or Material | 6-Chlorochroman-4-one |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'