missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%, Honeywell™

0.99

£38.96 - £316.58

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15635790
View Documents
Honeywell Research Chemicals
537705-1L
1 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15645790
View Documents
Honeywell Research Chemicals
537705-20L
20 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

110-12-3
114.188
FFWSICBKRCICMR-UHFFFAOYSA-N
8034
CC(C)CCC(=O)C
C7H14O
MFCD00008950
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
5-methylhexan-2-one

Specifications

110-12-3
C7H14O
MFCD00008950
506163
FFWSICBKRCICMR-UHFFFAOYSA-N
5-methylhexan-2-one
8034
99%
0.814g/mL at 25°C
(CH3)2CHCH2CH2COCH3
UN2302
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
CC(C)CCC(=O)C
114.188
114.19g/mol
5-Methyl-2-hexanone
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : 3 / PGIII

missing translation for 'rtecsNumber' : 203-737-8

SDS
Documents

Documents

Product Certifications