missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%, Thermo Scientific™

Product Code. 15448546
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116480-60-5
Molecular Formula C15H15N3
Molecular Weight (g/mol) 237.306
MDL Number MFCD08276951
InChI Key HNOKHIURIJOQKM-UHFFFAOYSA-N
Synonym 3-1-methyl-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole-5-carbonitrile, 3-1-methyl-3,6-dihydro-2h-pyridin-4-yl-1h-indole-5-carbonitrile, methyltetrahydropyridinylindolecarbonitrile, 3-1-methyl-4-1,2,5,6-tetrahydropyridyl indole-5-carbonitrile, 5-cyano-3-1-methyl-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, 3-1-methyl-1,2,3,6-tetrahydro-4-pyridyl-1h-indole-5-carbonitrile, 3-1-methyl-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole-5-carbonitrile
PubChem CID 14973216
IUPAC Name 3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indole-5-carbonitrile
SMILES CN1CCC(=CC1)C2=CNC3=C2C=C(C=C3)C#N

Specifications

Color White to Yellow
Quantity 5 g
UN Number UN3439
Beilstein 11193296
Formula Weight 237.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Cyano-3-(1-methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'