missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol, 97%, Thermo Scientific Chemicals

£34.50 - £381.90

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409762
View Documents
Thermo Scientific Alfa Aesar
A10817.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11419762
View Documents
Thermo Scientific Alfa Aesar
A10817.36
500 g
£112.56
500g
Please to purchase this item. Need a web account? Register with us today!
11429762
View Documents
Thermo Scientific Alfa Aesar
A10817.0E
2500 g
£381.90
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

92-69-3
170.211
YXVFYQXJAXKLAK-UHFFFAOYSA-N
7103
4-phenylphenol
C12H10O
MFCD00002347
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
CHEBI:34422
C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

92-69-3
305°C to 308°C
C12H10O
UN3077
14,7305
Soluble in water (0.7mg/ml at 20°C),ether,most common organic solvents,methanol,and chloroform.
C1=CC=C(C=C1)C2=CC=C(C=C2)O
170.211
CHEBI:34422
98%
164°C to 168°C
165°C (329°F)
MFCD00002347
1907452
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
YXVFYQXJAXKLAK-UHFFFAOYSA-N
4-phenylphenol
7103
170.21
4-Phenylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 202-179-2

missing translation for 'rtecsNumber' : DV5850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 4-Hydroxybiphenyl

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : (1,1-Biphenyl)-4-ol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only