missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.
Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.
Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 75°C to 78°C |
Boiling Point | 157°C (4 mmHg) |
Odor | Odorless |
Quantity | 1 g |
UN Number | UN3077 |
Merck Index | 14,4881 |
Solubility Information | Insoluble in water. Soluble in ethanol (25mg/ml),chloroform (1:1),ether (1:2),acetone (1:1.5),aqueous solutions of alkali hydroxides and carbonates,dichloromethane,methanol (50mg/ml),and ethyl acetate. |
Formula Weight | 206.29 |
Percent Purity | 99% |
Chemical Name or Material | 4-Isobutyl-alpha-methylphenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'