Learn More
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
£19.80 - £208.00
Chemical Identifiers
CAS | 1073-70-7 |
---|---|
Molecular Formula | C6H7ClN2·HCl |
Molecular Weight (g/mol) | 179.05 |
MDL Number | MFCD00012943 |
InChI Key | YQVZREHUWCCHHX-UHFFFAOYSA-N |
Synonym | 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride |
PubChem CID | 71600 |
IUPAC Name | (4-chlorophenyl)hydrazine;hydrochloride |
SMILES | C1=CC(=CC=C1NN)Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10425872
|
Thermo Scientific Acros
404950050 |
5 g |
£19.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10301301
|
Thermo Scientific Acros
404950250 |
25 g |
£62.11
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13408558
|
Thermo Scientific Acros
404951000 |
100 g |
£208.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1073-70-7 | |
179.05 | |
YQVZREHUWCCHHX-UHFFFAOYSA-N | |
71600 | |
C1=CC(=CC=C1NN)Cl.Cl |
C6H7ClN2·HCl | |
MFCD00012943 | |
4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
(4-chlorophenyl)hydrazine;hydrochloride |
Specifications
1073-70-7 | |
100.0 | |
Pink to White | |
Authentic | |
Glass bottle | |
NHNH2C6H4Cl·HCl | |
15,I,106 | |
YQVZREHUWCCHHX-UHFFFAOYSA-N | |
(4-chlorophenyl)hydrazine;hydrochloride | |
71600 | |
97% | |
4-Chlorophenylhydrazine hydrochloride, 97% |
96.0 | |
203.0°C | |
1% max. | |
96.0 to 104% (Argentometry) | |
C6H7ClN2·HCl | |
MFCD00012943 | |
4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
C1=CC(=CC=C1NN)Cl.Cl | |
179.05 | |
179.05 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-030-9
- 4-Chlorophenylhydrazine hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only