Learn More
3M™ Protective Coverall 4565
3M™ Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays and particles. 3M protective coverall 4565 is non-microporous to offers chemical protection with permeation tested resistance and is certified to EN 14126 infective agents.
Brand: 3M™ 7000089686
Code : 94
Additional Details : Weight : 0.01000kg
Description
- Type 4/5/6 Coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
- Made of bicomponent nonwoven with ultra-low linting PE laminate for a noticeably comfortable drape
- 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
- Taped seams across the suit for increased hold out and strength reducing the risk of tearing and exposure
- Protection against EN 14126 Biological agents
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out Til class 2