Learn More
3-Bromo-4-methylthiophene, 95%, Thermo Scientific Chemicals
55.90 EUR - 171.00 EUR
Chemical Identifiers
CAS | 30318-99-1 |
---|---|
Molecular Formula | C5H5BrS |
Molecular Weight (g/mol) | 177.06 |
MDL Number | MFCD00130084 |
InChI Key | MBUSOPVRLCFJCS-UHFFFAOYSA-N |
Synonym | 3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene |
PubChem CID | 2734935 |
IUPAC Name | 3-bromo-4-methylthiophene |
SMILES | CC1=CSC=C1Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10301213
|
Thermo Scientific Acros
379960010 |
1 g | Glass bottle |
55.90 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10217842
|
Thermo Scientific Acros
379960050 |
5 g | Glass bottle |
171.00 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30318-99-1 | |
177.06 | |
MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
2734935 | |
CC1=CSC=C1Br |
C5H5BrS | |
MFCD00130084 | |
3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
3-bromo-4-methylthiophene |
Specifications
30318-99-1 | |
100.0 | |
1.5840g/mL | |
78°C | |
94% min. (GC) | |
1.5770 to 1.579 | |
1.584 | |
MBUSOPVRLCFJCS-UHFFFAOYSA-N | |
3-bromo-4-methylthiophene | |
2734935 | |
95% | |
3-Bromo-4-methylthiophene |
94.0 | |
Colorless to Yellow | |
179°C to 183°C (770.0 mmHg) | |
Authentic | |
C5H5BrS | |
MFCD00130084 | |
3-bromo-4-methyl-thiophene, thiophene, 3-bromo-4-methyl, pubchem21832, acmc-209her, 3-bromo-4-methylthiophen, 4-bromo-3-methylthiophene, bidd:gt0486, 3-bromo-4-methylthiophene | |
CC1=CSC=C1Br | |
177.06 | |
177.06 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if
GHS Signal Word: Warning
- 3-Bromo-4-methylthiophene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only