missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15415385
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1241382-67-1
Molecular Formula C6H2BrF3Zn
MDL Number MFCD06200638
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 3,4,5-trifluorophenylzinc bromide, 3,4,5-trifluorophenylzinc bromide 0.5 m in tetrahydrofuran, 3,4,5-trifluorophenylzinc bromide, 0.5m in tetrahydrofuran, 3,4,5-trifluorophenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key PAKPKQOLJAUXJM-UHFFFAOYSA-M
SMILES [Zn+]Br.FC1=C[C-]=CC(F)=C1F
IUPAC Name bromozinc(1+);1,2,3-trifluorobenzene-5-ide
Molecular Weight (g/mol) 276.36
PubChem CID 24722635
Formula Weight 276.36
Chemical Name or Material 3,4,5-Trifluorophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'