Learn More
Chemical Identifiers
CAS | 135-19-3 |
---|---|
Molecular Formula | C10H8O |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00004067 |
InChI Key | JWAZRIHNYRIHIV-UHFFFAOYSA-N |
Synonym | 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol |
PubChem CID | 8663 |
ChEBI | CHEBI:10432 |
IUPAC Name | naphthalen-2-ol |
SMILES | C1=CC=C2C=C(C=CC2=C1)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10742801
|
Thermo Scientific Acros
180821000 |
100 g | Glass bottle |
£35.98
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752801
|
Thermo Scientific Acros
180825000 |
500 g | Glass bottle |
£98.49
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276753
|
Thermo Scientific Acros
180820010 |
1 kg | Glass bottle |
£178.89
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337813
|
Thermo Scientific Acros
180820025 |
2.5 kg | Plastic drum |
£387.93
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15159213
|
Thermo Scientific Acros
180820100 |
10 kg | Plastic drum | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
135-19-3 | |
144.17 | |
JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
8663 | |
naphthalen-2-ol |
C10H8O | |
MFCD00004067 | |
2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
CHEBI:10432 | |
C1=CC=C2C=C(C=CC2=C1)O |
Specifications
135-19-3 | |
100.0 | |
White | |
153°C | |
99% min. (GC) | |
C10H7OH | |
06,627 | |
2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
naphthalen-2-ol | |
8663 | |
144.17 | |
Crystals, Powder or Granules |
99.0 | |
120.0°C to 124.0°C | |
285.0°C to 286.0°C | |
Authentic | |
C10H8O | |
MFCD00004067 | |
15,6469 | |
Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol | |
C1=CC=C2C=C(C=CC2=C1)O | |
144.17 | |
CHEBI:10432 | |
99+% | |
2-Naphthol, 99+% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-182-7
missing translation for 'rtecsNumber' : QL2975000
missing translation for 'tsca' : TSCA
- 2-Naphthol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only