missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylbiphenyl-2-carboxaldehyde, 98%, Thermo Scientific™

£53.60 - £192.00

Chemical Identifiers

CAS 7111-68-4
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD03424630
InChI Key SEEYJCMPEVOYTF-UHFFFAOYSA-N
Synonym 2-2-methylphenyl benzaldehyde, 2'-methylbiphenyl-2-carbaldehyde, 2'-methyl-1,1'-biphenyl-2-carbaldehyde, 2'-methyl-biphenyl-2-carboxaldehyde, 2'-methylbiphenyl-2-carboxaldehyde, toluol-benzaldehyd, 2-o-tolyl benzaldehyde, 2'-methyl-biphenyl-2-carbaldehyde, 2-methyl-1,1-biphenyl-2-carbaldehyde, 2'-methyl 1,1'-biphenyl-2-carbaldehyde
PubChem CID 1393189
IUPAC Name 2-(2-methylphenyl)benzaldehyde
SMILES CC1=CC=CC=C1C1=CC=CC=C1C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479420
View Documents
Thermo Scientific Alfa Aesar
H50285.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11489420
View Documents
Thermo Scientific Alfa Aesar
H50285.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7111-68-4
196.25
SEEYJCMPEVOYTF-UHFFFAOYSA-N
1393189
CC1=CC=CC=C1C1=CC=CC=C1C=O
C14H12O
MFCD03424630
2-2-methylphenyl benzaldehyde, 2'-methylbiphenyl-2-carbaldehyde, 2'-methyl-1,1'-biphenyl-2-carbaldehyde, 2'-methyl-biphenyl-2-carboxaldehyde, 2'-methylbiphenyl-2-carboxaldehyde, toluol-benzaldehyd, 2-o-tolyl benzaldehyde, 2'-methyl-biphenyl-2-carbaldehyde, 2-methyl-1,1-biphenyl-2-carbaldehyde, 2'-methyl 1,1'-biphenyl-2-carbaldehyde
2-(2-methylphenyl)benzaldehyde

Specifications

7111-68-4
MFCD03424630
2-2-methylphenyl benzaldehyde, 2'-methylbiphenyl-2-carbaldehyde, 2'-methyl-1,1'-biphenyl-2-carbaldehyde, 2'-methyl-biphenyl-2-carboxaldehyde, 2'-methylbiphenyl-2-carboxaldehyde, toluol-benzaldehyd, 2-o-tolyl benzaldehyde, 2'-methyl-biphenyl-2-carbaldehyde, 2-methyl-1,1-biphenyl-2-carbaldehyde, 2'-methyl 1,1'-biphenyl-2-carbaldehyde
CC1=CC=CC=C1C1=CC=CC=C1C=O
196.25
196.25
2'-Methylbiphenyl-2-carboxaldehyde
C14H12O
Air Sensitive
SEEYJCMPEVOYTF-UHFFFAOYSA-N
2-(2-methylphenyl)benzaldehyde
1393189
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications