missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanthraquinone, 98%, Thermo Scientific Chemicals

Product Code. 11327186
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-51-5
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00001237
InChI Key SJEBAWHUJDUKQK-UHFFFAOYSA-N
Synonym 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
PubChem CID 6772
IUPAC Name 2-ethylanthracene-9,10-dione
SMILES CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 107°C to 111°C
Boiling Point 180°C to 190°C
Flash Point >210°C (410°F)
Quantity 100 g
Beilstein 1969873
Solubility Information Insoluble in water.
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material 2-Ethylanthraquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'